Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199949.2 | NP_956243.1 | 5 - 6 | 811 | 90 | |
GenBank mRNA | AY398339.1 | - | 5 - 6 | 811 | 90 | |
BC046042.1 | - | 5 - 6 | 594 | 90 | ||
BC071464.1 | - | 5 - 6 | 596 | 90 | ||
BC150470.1 | - | 5 - 6 | 595 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |