Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl6aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200734.1 | NP_957028.1 | 8 - 9 | 2023 | 69 | |
XM_017356456.2 | XP_017211945.1 | 8 - 9 | 2055 | 69 | ||
XM_021476813.1 | XP_021332488.1 | 9 - 10 | 2143 | 69 | ||
XM_021476814.1 | XP_021332489.1 | 8 - 9 | 2009 | 69 | ||
XM_021476815.1 | XP_021332490.1 | 8 - 9 | 2318 | 69 | ||
GenBank mRNA | BC059515.1 | - | 8 - 9 | 2023 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |