Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rasa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099451.1 | NP_001092921.1 | 18 - 19 | 2101 | 63 | |
XM_005171995.3 | XP_005172052.1 | 18 - 19 | 2351 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Danio rerio Probe spans exons |