Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csf2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113733.1 | NP_001107205.1 | 7 - 8 | 948 | 83 | |
XM_005171550.4 | XP_005171607.1 | 7 - 8 | 1057 | 83 | ||
XM_021469067.1 | XP_021324742.1 | 7 - 8 | 1057 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |