Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114552.2 | NP_001108024.1 | 1 - 2 | 436 | 96 | |
GenBank mRNA | EU203154.1 | - | 1 - 2 | 437 | 96 | |
FJ915144.1 | - | 1 - 2 | 142 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |