Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114925.1 | NP_001108397.1 | 3 - 4 | 698 | 131 | |
XM_005170161.4 | XP_005170218.1 | 4 - 5 | 131 | |||
GenBank mRNA | BC155622.1 | - | 3 - 4 | 698 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |