Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115064.2 | NP_001108536.2 | 4 - 5 | 481 | 76 | |
GenBank mRNA | EF137867.1 | - | 4 - 5 | 481 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |