Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139480.1 | NP_001132952.1 | 1 - 2 | 600 | 67 | |
XM_005173662.4 | XP_005173719.1 | 1 - 2 | 614 | 67 | ||
GenBank mRNA | BC129216.1 | - | 1 - 2 | 569 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |