Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpp6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115122.1 | NP_001108594.1 | 15 - 16 | 1615 | 73 | |
XM_005163289.3 | XP_005163346.1 | 10 - 11 | 1583 | 73 | ||
XM_005163290.4 | XP_005163347.1 | 10 - 11 | 1058 | 73 | ||
XM_005163291.4 | XP_005163348.1 | 10 - 11 | 1026 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |