Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp4r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115140.1 | NP_001108612.1 | 3 - 4 | 417 | 97 | |
GenBank mRNA | BC060912.1 | - | 3 - 4 | 355 | 97 | |
BC151885.1 | - | 3 - 4 | 376 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |