Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001118896.2 | NP_001112368.2 | 5 - 6 | 625 | 75 | |
XM_005162680.4 | XP_005162737.1 | 5 - 6 | 604 | 75 | ||
GenBank mRNA | BC045519.1 | - | 4 - 5 | 550 | 75 | |
EH559864.1 | - | 2 - 3 | 256 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |