Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001118896.2 | NP_001112368.2 | 8 - 9 | 1089 | 85 | |
XM_005162680.4 | XP_005162737.1 | 8 - 9 | 1050 | 85 | ||
GenBank mRNA | BC045519.1 | - | 7 - 8 | 1014 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |