Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122836.1 | NP_001116308.1 | 3 - 4 | 414 | 122 | |
GenBank mRNA | BC139583.1 | - | 3 - 4 | 381 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |