Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122874.1 | NP_001116346.1 | 4 - 5 | 1545 | 72 | |
XM_005160320.4 | XP_005160377.1 | 4 - 5 | 1539 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |