Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131036.2 | NP_571111.2 | 11 - 12 | 2405 | 115 | |
XM_005171264.4 | XP_005171321.1 | 12 - 13 | 2664 | 115 | ||
XM_005171265.3 | XP_005171322.1 | 10 - 11 | 2136 | 115 | ||
XM_009298633.3 | XP_009296908.1 | 12 - 13 | 2400 | 115 | ||
GenBank mRNA | AJ001423.1 | - | 11 - 12 | 2365 | 115 | |
BC095389.1 | - | 10 - 11 | 2064 | 115 | ||
KM655642.1 | - | 10 - 11 | 2239 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |