Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200952.2 | NP_957246.1 | 2 - 3 | 333 | 87 | |
GenBank mRNA | AY394941.1 | - | 2 - 3 | 333 | 87 | |
BC053288.1 | - | 2 - 3 | 299 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |