Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320519.1 | NP_001307448.1 | 4 - 5 | 605 | 77 | |
XM_017354980.2 | XP_017210469.1 | 4 - 5 | 648 | 77 | ||
XM_017354981.2 | XP_017210470.1 | 4 - 5 | 728 | 77 | ||
GenBank mRNA | BC053185.1 | - | 4 - 5 | 838 | 77 | |
BC165151.1 | - | 3 - 4 | 536 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |