Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201050.1 | NP_957344.1 | 5 - 6 | 776 | 122 | |
GenBank mRNA | BC048055.1 | - | 5 - 6 | 776 | 122 | |
BC164531.1 | - | 4 - 5 | 674 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |