Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327787.1 | NP_001314716.1 | 12 - 13 | 2218 | 104 | |
NR_137174.1 | 13 - 14 | 2403 | 104 | |||
XM_005164196.4 | XP_005164253.1 | 12 - 13 | 2227 | 104 | ||
XM_005164197.4 | XP_005164254.1 | 12 - 13 | 2224 | 104 | ||
XM_005164198.4 | XP_005164255.1 | 12 - 13 | 2221 | 104 | ||
GenBank mRNA | BC052767.1 | - | 13 - 14 | 2305 | 104 | |
HM151009.1 | - | 12 - 13 | 1980 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |