Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327787.1 | NP_001314716.1 | 16 - 17 | 2558 | 117 | |
NR_137174.1 | 17 - 18 | 2743 | 117 | |||
XM_005164196.4 | XP_005164253.1 | 16 - 17 | 2567 | 117 | ||
XM_005164197.4 | XP_005164254.1 | 16 - 17 | 2564 | 117 | ||
XM_005164198.4 | XP_005164255.1 | 16 - 17 | 2561 | 117 | ||
GenBank mRNA | BC052767.1 | - | 17 - 18 | 2645 | 117 | |
HM151009.1 | - | 16 - 17 | 2320 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |