Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bicc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203420.1 | NP_981965.1 | 18 - 19 | 2447 | 67 | |
GenBank mRNA | AY513238.1 | - | 18 - 19 | 2447 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Danio rerio Probe spans exons |