Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bicc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203420.1 | NP_981965.1 | 5 - 6 | 600 | 69 | |
GenBank mRNA | AY513238.1 | - | 5 - 6 | 600 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |