Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212636.1 | NP_997801.1 | 6 - 7 | 618 | 126 | |
GenBank mRNA | BC054945.1 | - | 6 - 7 | 549 | 126 | |
BC066574.1 | - | 6 - 7 | 618 | 126 | ||
BC155178.1 | - | 6 - 7 | 680 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |