Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213034.3 | NP_998199.1 | 2 - 3 | 267 | 96 | |
GenBank mRNA | BC059677.1 | - | 2 - 3 | 91 | 96 | |
EH440189.1 | - | 2 - 3 | 131 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |