Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213101.1 | NP_998266.1 | 3 - 4 | 589 | 65 | |
GenBank mRNA | BC068325.1 | - | 3 - 4 | 589 | 65 | |
CU458908.1 | - | 3 - 4 | 589 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |