Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213352.2 | NP_998517.2 | 1 - 2 | 157 | 107 | |
GenBank mRNA | BC057417.1 | - | 1 - 2 | 111 | 104 | |
BC152216.1 | - | 1 - 2 | 104 | 101 | ||
CB923490.1 | - | 1 - 2 | 57 | 107 | ||
EH449789.1 | - | 1 - 2 | 136 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |