Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213451.2 | NP_998616.2 | 5 - 6 | 581 | 84 | |
XM_005163280.4 | XP_005163337.1 | 5 - 6 | 658 | 84 | ||
GenBank mRNA | BC055242.1 | - | 5 - 6 | 84 | ||
DT067300.1 | - | 1 - 2 | 161 | 84 | ||
DT072464.1 | - | 5 - 6 | 518 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |