Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vcanb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214688.1 | NP_999853.1 | 2 - 3 | 105 | 81 | |
GenBank mRNA | AB119257.1 | - | 2 - 3 | 105 | 81 | |
BC162605.1 | - | 2 - 3 | 91 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |