Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC070010.1 | - | 4 | 1720 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |