Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081760.1 | NP_001075229.1 | 36 - 37 | 5605 | 79 | |
XM_023651654.1 | XP_023507422.1 | 5719 | 79 | |||
XM_023651655.1 | XP_023507423.1 | 5701 | 79 | |||
GenBank mRNA | AB088365.1 | - | 36 - 37 | 5605 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |