Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081786.1 | NP_001075255.1 | 3 - 4 | 327 | 141 | |
GenBank mRNA | DQ385611.1 | - | 3 - 4 | 327 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |