Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081807.2 | NP_001075276.1 | 9 - 10 | 684 | 73 | |
XM_023644108.1 | XP_023499876.1 | 728 | 73 | |||
XM_023644109.1 | XP_023499877.1 | 737 | 73 | |||
XM_023644110.1 | XP_023499878.1 | 674 | 73 | |||
XM_023644111.1 | XP_023499879.1 | 620 | 73 | |||
XM_023644112.1 | XP_023499880.1 | 617 | 73 | |||
GenBank mRNA | AF141931.1 | - | 9 - 10 | 692 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Equus caballus Probe spans exons |