Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081816.1 | NP_001075285.1 | 8 - 9 | 1160 | 88 | |
XM_005599763.3 | XP_005599820.2 | 1326 | 88 | |||
XM_005599765.3 | XP_005599822.1 | 1222 | 88 | |||
XM_023618377.1 | XP_023474145.1 | 1207 | 88 | |||
GenBank mRNA | AB033415.1 | - | 8 - 9 | 1160 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Equus caballus Inventoried Probe spans exons |