Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081818.1 | NP_001075287.1 | 2 - 3 | 1093 | 63 | |
XM_023626097.1 | XP_023481865.1 | 1149 | 63 | |||
XM_023626103.1 | XP_023481871.1 | 1149 | 63 | |||
GenBank mRNA | AB034731.1 | - | 2 - 3 | 1093 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Equus caballus Probe spans exons |