Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081919.1 | NP_001075388.1 | 1 | 1355 | 127 | |
XM_005600327.3 | XP_005600384.1 | 3134 | 127 | |||
XM_023619753.1 | XP_023475521.1 | 1786 | 127 | |||
XM_023619754.1 | XP_023475522.1 | 3104 | 127 | |||
XM_023619755.1 | XP_023475523.1 | 1517 | 127 | |||
XM_023619756.1 | XP_023475524.1 | 2051 | 127 | |||
XM_023619757.1 | XP_023475525.1 | 1744 | 127 | |||
XM_023619758.1 | XP_023475526.1 | 1762 | 127 | |||
XM_023619759.1 | XP_023475527.1 | 1722 | 127 | |||
XM_023619760.1 | XP_023475528.1 | 1988 | 127 | |||
XM_023619761.1 | XP_023475529.1 | 1473 | 127 | |||
XM_023619762.1 | XP_023475530.1 | 2140 | 127 | |||
GenBank mRNA | DQ681103.1 | - | 1 | 1355 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |