Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082494.1 | NP_001075963.1 | 5 - 6 | 530 | 67 | |
GenBank mRNA | U60978.1 | - | 5 - 6 | 530 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Equus caballus Probe spans exons |