Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082510.2 | NP_001075979.2 | 1 - 2 | 237 | 85 | |
XM_023651597.1 | XP_023507365.1 | 590 | 85 | |||
GenBank mRNA | Y09440.1 | - | 254 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |