Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083951.2 | NP_001077420.2 | 3 - 4 | 381 | 79 | |
GenBank mRNA | AY184956.1 | - | 3 - 4 | 384 | 79 | |
CD536564.1 | - | 3 - 4 | 381 | 79 | ||
JL626338.1 | - | 3 - 4 | 312 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Inventoried Probe spans exons |