Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089586.1 | NP_001083055.1 | 6 - 7 | 2241 | 105 | |
GenBank mRNA | DQ497159.1 | - | 6 - 7 | 2241 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Equus caballus Probe spans exons |