Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163955.1 | NP_001157427.1 | 5 - 6 | 1148 | 101 | |
XM_023627692.1 | XP_023483460.1 | 1326 | 101 | |||
GenBank mRNA | AB043677.1 | - | 5 - 6 | 1115 | 101 | |
HQ890110.1 | - | 2 - 3 | 473 | 101 | ||
JL617688.1 | - | 3 - 4 | 600 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |