Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917787.5 | XP_001917822.2 | 759 | 91 | ||
GenBank mRNA | DQ492250.1 | - | 1 - 2 | 206 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Equus caballus Probe spans exons |