Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497744.2 | XP_001497794.1 | 1029 | 69 | ||
GenBank mRNA | EU029663.1 | - | 2 - 3 | 273 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |