Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DQB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142813.1 | NP_001136285.1 | 5 - 6 | 840 | 89 | |
GenBank mRNA | JQ254075.1 | - | 5 - 6 | 815 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Equus caballus Probe spans exons |