Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081865.1 | NP_001075334.1 | 3 | 352 | 75 | |
GenBank mRNA | AF506973.1 | - | 3 | 352 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |