Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159692.1 | NP_001153164.1 | 4 - 5 | 906 | 67 | |
GenBank mRNA | FJ028654.1 | - | 4 - 5 | 906 | 67 | |
JF749865.1 | - | 4 - 5 | 890 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |