Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTHLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163981.1 | NP_001157453.1 | 4 - 5 | 863 | 93 | |
XM_005610810.3 | XP_005610867.1 | 873 | 93 | |||
XM_014740461.2 | XP_014595947.1 | 1011 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Equus caballus Probe spans exons |