Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205192.1 | NP_001192121.1 | 6 - 7 | 1023 | 70 | |
XM_023644173.1 | XP_023499941.1 | 1564 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Equus caballus Probe spans exons |