Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205261.1 | NP_001192190.1 | 7 - 8 | 1643 | 87 | |
XM_014730313.2 | XP_014585799.2 | 1632 | 87 | |||
XM_014730314.2 | XP_014585800.2 | 1608 | 87 | |||
XM_014730317.2 | XP_014585803.2 | 1482 | 87 | |||
GenBank mRNA | DN506315.1 | - | 1 - 2 | 104 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |