Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 8 - 9 | 265 | 83 | |
XM_014738515.2 | XP_014594001.1 | 276 | 134 | |||
GenBank mRNA | GU196267.1 | - | 8 - 9 | 265 | 83 | |
KP658381.1 | - | 9 - 10 | 272 | 134 | ||
KP658382.1 | - | 7 - 8 | 221 | 83 | ||
KT368778.1 | - | 9 - 10 | 266 | 134 | ||
KT368779.1 | - | 7 - 8 | 215 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |