Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032896.1 | 1 | 57 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox B3 | HOXB3 | -4.5kb | Sense | Chr. 11 - 24672870 - 24699039 on NCBI Build EquCab3.0 | |
Downstream | homeobox B5 | HOXB5 | 11.3kb | Sense | Chr. 11 - 24714926 - 24717500 on NCBI Build EquCab3.0 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Equus caballus Both primers and probe map within a single exon |